missing translation for 'onlineSavingsMsg'
Learn More

m-Toluoyl chloride, 99%

Product Code. 10367230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367230 100 g Glass bottle 100g
10784081 5 g Glass bottle 5g
10163630 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367230 Supplier Thermo Scientific Acros Supplier No. 153411000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1711-06-4 | C8H7ClO | 154.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1711-06-4
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000681
InChI Key YHOYYHYBFSYOSQ-UHFFFAOYSA-N
Synonym m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
PubChem CID 74375
IUPAC Name 3-methylbenzoyl chloride
SMILES CC1=CC=CC(=C1)C(=O)Cl

Tekniske data

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 1.1700g/mL
Boiling Point 86.0°C (5.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4COCl
Refractive Index 1.5475 to 1.5495
Quantity 100 g
Beilstein 09, 477
Specific Gravity 1.17
Solubility Information Solubility in water: decomposes.
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Toluoyl chloride, 99%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • m-Toluoyl chloride
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.