missing translation for 'onlineSavingsMsg'
Learn More

m-Toluic acid, 99%

Codice prodotto. 11444854
Cambia vista
Click to view available options
Quantity:
250 g
1000 g
5000 g
Dimensione della confezione:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
11444854 5000 g 5000g
11434854 250 g 250g
11424854 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11444854 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A13785.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 99-04-7 | C8H8O2 | 136.15 g/mol

m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

Solubility
Soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 99-04-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002523
InChI Key GPSDUZXPYCFOSQ-UHFFFAOYSA-N
Synonym m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
PubChem CID 7418
ChEBI CHEBI:10589
IUPAC Name 3-methylbenzoic acid
SMILES CC1=CC=CC(=C1)C(O)=O

Specifica

Melting Point 108°C to 110°C
Density 1.05
Boiling Point 263°C
Flash Point 150°C (302°F)
Quantity 5000 g
Beilstein 970526
Merck Index 14,9535
Solubility Information Soluble in alcohol and ether.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material m-Toluic acid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.