missing translation for 'onlineSavingsMsg'
Learn More

m-Toluic acid, 99%, Thermo Scientific Chemicals

Product Code. 10573111
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-04-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002523
InChI Key GPSDUZXPYCFOSQ-UHFFFAOYSA-N
Synonym m-toluic acid, m-toluylic acid, 3-toluic acid, benzoic acid, 3-methyl, meta-toluic acid, m-methylbenzoic acid, beta-methylbenzoic acid, 3-methyl-benzoic acid, beta-bethylbenzoic acid, unii-1ua7k8eext
PubChem CID 7418
ChEBI CHEBI:10589
IUPAC Name 3-methylbenzoic acid
SMILES CC1=CC=CC(=C1)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 108°C to 112°C
Color Beige-Yellow to Yellow
Density 1.05
Boiling Point 263°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3C6H4CO2H
Quantity 5 g
Beilstein 09, 475
Merck Index 14, 9535
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 136.15
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material m-Toluic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'