missing translation for 'onlineSavingsMsg'
Learn More

m-Toluenesulfonyl chloride, 98%

Product Code. 11462817
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462817 25 g 25g
11428790 1 g 1g
11452817 5 g 5g
11472817 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11462817 Supplier Thermo Scientific Alfa Aesar Supplier No. A15458.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1899-93-0 | C7H7ClO2S | 190.641 g/mol

Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant involved in they synthesis of biologically active molecules including small molecule ligands for the Stat3 SH2 domain, aryldisulfonamides with antibacterial activity, quinazoline analogs for the treatment of Gaucher disease, 17β-HSD2 inhibitors for the treatment of osteoporosis, isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1899-93-0
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00051747
InChI Key KFPMLWUKHQMEBU-UHFFFAOYSA-N
Synonym m-toluenesulfonyl chloride, 3-methylbenzene-1-sulfonyl chloride, 3-toluenesulfonyl chloride, 3-methyl-benzenesulfonyl chloride, 3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-methyl, 3-toluenesulphonyl chloride, 3-methylphenylsulfonyl chloride, pubchem5624, m-tolylsulfonyl chloride
PubChem CID 2734595
IUPAC Name 3-methylbenzenesulfonyl chloride
SMILES CC1=CC(=CC=C1)S(=O)(=O)Cl

Specifications

Density 1.314
Boiling Point 252°C to 253°C
Flash Point 107°C (224°F)
Odor Acrid
Refractive Index 1.548
Quantity 25 g
UN Number UN3265
Beilstein 2087865
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 190.65
Percent Purity 98%
Chemical Name or Material m-Toluenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.