missing translation for 'onlineSavingsMsg'
Learn More

m-Toluenesulfonic acid monohydrate, 97%

Product Code. 11352146
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352146 5 g 5g
11362146 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352146 Supplier Thermo Scientific Alfa Aesar Supplier No. L03419.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 312619-56-0 | C7H10O4S | 190.213 g/mol

m-Toluenesulfonic acid monohydrate is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluenesulfonic acid monohydrate is used in chemical research.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from strong bases, moisture. Incompatible materials are oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312619-56-0
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00167130
InChI Key GHLYGGIKYFLYTH-UHFFFAOYSA-N
Synonym 3-methylbenzenesulfonic acid hydrate, m-toluenesulfonic acid monohydrate, m-toluenesulfonic acid hydrate, acmc-20akdl, m-toluenesulfonic acid 1-hydrate, 3-toluenesulfonic acid 1-hydrate, 3-methylbenzenesulfonic acid hydrate 1:x, benzenesulfonicacid, 3-methyl-, hydrate 1:1
PubChem CID 53249652
IUPAC Name 3-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC(=CC=C1)S(=O)(=O)O.O

Specifications

Boiling Point 165°C (0.1 mmHg)
Refractive Index 1.527
Quantity 5 g
UN Number UN2586
Beilstein 2208421
Solubility Information Fully miscible in water.
Formula Weight 190.22 (172.20 Anhydrous)
Percent Purity 97%
Chemical Name or Material m-Toluenesulfonic acid monohydrate
compliance-icons
Product Identifier
  • m-Toluenesulfonic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: m-Toluenesulfonic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.