missing translation for 'onlineSavingsMsg'
Learn More

m-Tolualdehyde, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-200978
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
620-23-5, 123-31-9
C8H8O
120.15
MFCD00003374
OVWYEQOVUDKZNU-UHFFFAOYSA-N
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
12105
CHEBI:28476
3-methylbenzaldehyde
CC1=CC=CC(=C1)C=O
This item is not returnable. View return policy
620-23-5, 123-31-9
C8H8O
120.15
MFCD00003374
OVWYEQOVUDKZNU-UHFFFAOYSA-N
m-tolualdehyde, benzaldehyde, 3-methyl, m-methylbenzaldehyde, 3-tolylaldehyde, 3-tolualdehyde, m-tolyl aldehyde, m-toluylaldehyde, 3-methyl-benzaldehyd, unii-owh6650c4y, 3-methyl benzaldehyde
12105
CHEBI:28476
3-methylbenzaldehyde
CC1=CC=CC(=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.0190g/mL
Boiling Point 199°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CHO
Refractive Index 1.5400 to 1.542
Quantity 25 g
Beilstein 07, 296
Specific Gravity 1.019
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
Stabilizer 0.1% hydroquinone
Formula Weight 120.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material m-Tolualdehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • m-Tolualdehyde, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'