missing translation for 'onlineSavingsMsg'
Learn More

m-Salicylic Acid, TRC

Product Code. 30138103
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138103 50 g 50g
17962735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138103 Supplier Toronto Research Chemicals Supplier No. TRCS08810550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7 H6 O3
Molecular Weight (g/mol) 138.12
Synonym Benzoic acid, 3-hydroxy-, Benzoic acid, m-hydroxy- (8CI), 3-Hydroxybenzoic acid, 3-Carboxyphenol, NSC 55746, m-Carboxyphenol, m-Hydroxybenzoic acid, m-Salicylic acid
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)c1cccc(O)c1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H6O3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H,(H,9,10)
Formula Weight 138.0317
Chemical Name or Material 3-Hydroxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.