missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-Ras/R-Ras3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | M-Ras/R-Ras3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313074
|
Novus Biologicals
NBP3-03804-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18361056
|
Novus Biologicals
NBP3-03804-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
M-Ras/R-Ras3 Polyclonal specifically detects M-Ras/R-Ras3 in Mouse samples. It is validated for Western Blot.Specifications
| M-Ras/R-Ras3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 79-208 of human M-Ras/R-Ras3 (NP_036351.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 22808 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title