missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-Ras/R-Ras3 Antibody (OTI3C4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72776CL1
This item is not returnable.
View return policy
Description
M-Ras/R-Ras3 Monoclonal antibody specifically detects M-Ras/R-Ras3 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| M-Ras/R-Ras3 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
| Full length human recombinant protein of human MRAS (NP_036351) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 22808 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI3C4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction