Learn More
Julabo™ M+R In-Line Pt100 Sensor
Product Code. 15244353
Change view
Click to view available options
Connection Type:
2 - M24 x 1.5 Male Connections
2 - M30 x 1.5 Male Connections
2 - M38 x 1.5 Male Connections
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type unitSize
15244353 2 - M38 x 1.5 Male Connections Each
15214353 2 - M24 x 1.5 Male Connections Each
15224353 2 - M30 x 1.5 Male Connections Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15244353 Supplier Julabo™ Supplier No. 8981023
Specifications
Description M+R In-Line Pt100 Sensor
For Use With (Equipment) Suitable for PRESTO W91, PRESTO W91t, PRESTO W91tt, PRESTO W91ttx Temperature Control System
Connection Type 2 - M38 x 1.5 Male Connections