Learn More
Julabo™ M+R In-Line Pt100 Sensor
Click to view available options
Connection Type:
2 - M24 x 1.5 Male Connections
2 - M30 x 1.5 Male Connections
2 - M38 x 1.5 Male Connections
Unit Size:
Each
Specifications
Specifications
| Description | M+R In-Line Pt100 Sensor |
| For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System |
| Connection Type | 2 - M24 x 1.5 Male Connections |