missing translation for 'onlineSavingsMsg'
Learn More

m-Phenylenediamine, 98%

Product Code. p-7022519
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11408256

Brand: Thermo Scientific Alfa Aesar A15339.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-45-2 | C6H8N2 | 108.144 g/mol

m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Phenylenediamine is used in the foaming-type hair dye composition. It is used as a synthetic electronic grade polyimide material and epoxy resin curing agent. It is used also block polymers, textile fibers, urethanes, petroleum additives, rubber chemicals, in corrosion inhibitors, in photography, as reagent for gold & bromine.

Solubility
Soluble in water, methanol, acetone, DMF, MEK, dioxane

Notes
Air & Light Sensitive. Store in the dark. Store under dry argon. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-45-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007799
InChI Key WZCQRUWWHSTZEM-UHFFFAOYSA-N
Synonym m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine
PubChem CID 7935
ChEBI CHEBI:8092
IUPAC Name benzene-1,3-diamine
SMILES C1=CC(=CC(=C1)N)N

Specifications

Melting Point 62°C to 66°C
Density 1.139
Boiling Point 284°C to 287°C
Flash Point 187°C (368°F)
Quantity 250 g
UN Number UN1673
Beilstein 471357
Sensitivity Air and light sensitive
Merck Index 14,7283
Solubility Information Soluble in water, methanol, acetone, DMF, MEK, dioxane
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material m-Phenylenediamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.