missing translation for 'onlineSavingsMsg'
Learn More

m,p-Quaterphenyl, TRC

Product Code. 30423098
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423098 50 mg
30409610 250 mg
30422675 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423098 Supplier Toronto Research Chemicals Supplier No. TRCQ11655050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1166-19-4
Molecular Formula C24 H18
Molecular Weight (g/mol) 306.4
Synonym 3,4'-Diphenylbiphenyl, 1,1':3',1' ':4' ',1' ' '-Quaterphenyl
IUPAC Name 1-phenyl-3-(4-phenylphenyl)benzene
SMILES c1ccc(cc1)c2ccc(cc2)c3cccc(c3)c4ccccc4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H18/c1-3-8-19(9-4-1)21-14-16-22(17-15-21)24-13-7-12-23(18-24)20-10-5-2-6-11-20/h1-18H
Formula Weight 306.141
Chemical Name or Material m,p-Quaterphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.