missing translation for 'onlineSavingsMsg'
Learn More

m-Nitrophenol, 99.9%, For GC analysis, MP Biomedicals™

Product Code. 11409181 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409181 5 g 5g
11419181 25 g 25g
11429181 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409181 Supplier MP Biomedicals Supplier No. 0215589305
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES OC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Color Red-Yellow
Quantity 5 g
Formula Weight 139.11
Percent Purity 0.999
Physical Form Crystalline Powder
Chemical Name or Material m-Nitrophenol, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.