missing translation for 'onlineSavingsMsg'
Learn More

m-Nitroacetophenone, 98%

Product Code. 10336630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336630 100 g Plastic bottle 100g
10503681 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336630 Supplier Thermo Scientific Acros Supplier No. 128331000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-89-1 | C8H7NO3 | 165.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76.0°C to 79.0°C
Boiling Point 201.0°C to 203.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4COCH3
Quantity 100 g
Beilstein 07, 288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 165.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material m-Nitroacetophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.