missing translation for 'onlineSavingsMsg'
Learn More

m-Isopropoxyaniline, TRC

Product Code. 30393752
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393752 5 g
30402581 25 g
30399697 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393752 Supplier Toronto Research Chemicals Supplier No. TRCI8229905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41406-00-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
Synonym 3-(Isopropyloxy)aniline, 3-Isopropoxyaniline, 3-Isopropoxybenzenamine, m-(Isopropyloxy)aniline, m-Aminophenyl Isopropyl Ether
IUPAC Name 3-propan-2-yloxyaniline
SMILES CC(C)Oc1cccc(N)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H13NO/c1-7(2)11-9-5-3-4-8(10)6-9/h3-7H,10H2,1-2H3
Formula Weight 151.0997
Chemical Name or Material m-Isopropoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.