missing translation for 'onlineSavingsMsg'
Learn More

m-Hydroxy Mexiletine, TRC

Product Code. 30393622
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393622 25 mg -
30396098 5 mg 5mg
30398312 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393622 Supplier Toronto Research Chemicals Supplier No. TRCH93903025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133883-48-4
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.26
Synonym 3-(2-Aminopropoxy)-2,4-dimethylphenol
IUPAC Name 3-(2-aminopropoxy)-2,4-dimethylphenol
SMILES CC(N)COc1c(C)ccc(O)c1C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H17NO2/c1-7-4-5-10(13)9(3)11(7)14-6-8(2)12/h4-5,8,13H,6,12H2,1-3H3
Formula Weight 195.1259
Chemical Name or Material m-Hydroxy Mexiletine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.