missing translation for 'onlineSavingsMsg'
Learn More

m-Cymene, TRC

Product Code. 30357780
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357780 100 mg -
30354494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357780 Supplier Toronto Research Chemicals Supplier No. TRCC989765100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-77-3
Molecular Formula C10 H14
Molecular Weight (g/mol) 134.218
Synonym Benzene, 1-methyl-3-(1-methylethyl)-, m-Cymene (8CI), 1-Methyl-3-(1-methylethyl)benzene, 1-Isopropyl-3-methylbenzene, 1-Methyl-3-isopropylbenzene, 3-Isopropyltoluene, Meta-cymene, NSC 73975, m-Cymol, m-Isopropyltoluene, m-Methylisopropylbenzene, β-Cymene
IUPAC Name 1-methyl-3-propan-2-ylbenzene
SMILES CC(C)c1cccc(C)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H14/c1-8(2)10-6-4-5-9(3)7-10/h4-8H,1-3H3
Formula Weight 134.11
Chemical Name or Material 3-Isopropyltoluene (m-Cymol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.