missing translation for 'onlineSavingsMsg'
Learn More

M-CSFR/CD115 Antibody (61715) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30734778 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734778 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734778 Supplier Novus Biologicals Supplier No. FAB3292MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Human samples. It is validated for ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen M-CSFR/CD115
Applications ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 61715
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115, Ile20-Glu512 (Pro54Ala), Accession # P07333.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1436
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.