missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-CSFR/CD115 Antibody (460615), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3818CL1
This item is not returnable.
View return policy
Description
M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| M-CSFR/CD115 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
| Mouse myeloma cell line NS0-derived recombinant mouse M-CSF R/CD115, Ala20-Ser511, Accession # P09581 | |
| 0.1 mL | |
| Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase | |
| 1436 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 460615 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction