missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M-CSF Monoclonal Antibody (131621)

Rat Monoclonal Antibody
£444.00 - £576.00
Specifications
| Antigen | M-CSF |
|---|---|
| Clone | 131621 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15563462
|
Invitrogen™
MA523773 |
500 μg |
£576.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15563552
![]() |
Invitrogen™
MA523863 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In sandwich ELISAs, no cross-reactivity with recombinant human M-CSF, recombinant mouse (rm) SCF, rmFlt-3 Ligand, rmG-CSF, and rmGM-CSF is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.Specifications
| M-CSF | |
| 0.5 mg/mL | |
| ELISA, Neutralization | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 5% trehalose and No Preservative | |
| P07141 | |
| 12977 | |
| E. coli-derived recombinant mouse M-CSF Lys33-Glu262 (predicted). | |
| Antibody |
| 131621 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Lyophilized | |
| IgG2b | |
| Mouse | |
| Csf1 | |
| C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1 | |
| Csf1 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title