missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, TRC

Product Code. 30133220
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133220 250 g 250g
18053350 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133220 Supplier Toronto Research Chemicals Supplier No. TRCC781915250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7 H8 O
Molecular Weight (g/mol) 108.14
Synonym Metacresol, 3-Methylphenol, Phenol, 3-methyl- (9CI, ACI), 3-Methylphenol (ACI), m-Cresol (8CI), 1-Hydroxy-3-methylbenzene, 3-Cresol, 3-Hydroxytoluene, m-Cresylic acid, m-Hydroxytoluene, m-Kresol, m-Methylphenol, m-Oxytoluene, m-Toluol, meta-Cresol, NSC 8768
IUPAC Name 3-methylphenol
SMILES Cc1cccc(O)c1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3
Formula Weight 108.0575
Percent Purity >95%
Chemical Name or Material 3-Methylphenol (m-Cresol)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.