missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol Purple

Product Code. 11445767
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445767 5 g 5g
11455767 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445767 Supplier Thermo Scientific Alfa Aesar Supplier No. B24338.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2303-01-7 | C21H18O5S | 382.43 g/mol

m-Cresol Purple acts as a pH indicator dye. It plays an important role in spectrophotometric pH measurements. It operates in the pH ranges 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet). Further, it is employed to stain epithelial cells and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Cresol Purple acts as a pH indicator dye. It plays an important role in spectrophotometric pH measurements. It operates in the pH ranges 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet). Further, it is employed to stain epithelial cells and proteins.

Solubility
Soluble in water and sodium hydroxide solution. Slightly soluble in alcohol.

Notes
Transition interval (acid): pH 1.2 (red) to pH 2.8 (yellow); (alkaline) pH 7.4 (yellow) to pH 9.0 (purple)Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2303-01-7
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005871
InChI Key OLQIKGSZDTXODA-UHFFFAOYSA-N
Synonym m-Cresolsulfonephthalein
PubChem CID 73030
IUPAC Name 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C

Specifications

Beilstein 350314
Merck Index 14,2577
Solubility Information Soluble in water and sodium hydroxide solution. Slightly soluble in alcohol.
Formula Weight 382.44
Quantity 5 g
Chemical Name or Material m-Cresol Purple

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.