missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, 99%

Product Code. 10248223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Unit Size:
100g
10kg
1kg
2.5kg
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10248223 2.5 kg Glass bottle 2.5kg
10367523 100 g Glass bottle 100g
10204570 500 g Glass bottle 500g
10785171 1 kg Glass bottle 1kg
11984661 10 kg Metal Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10248223 Supplier Thermo Scientific Acros Supplier No. 110580025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 108-39-4 | C7H8O | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O

Specifications

Melting Point 8.0°C to 10.0°C
Color Colorless to Red
Density 1.0300g/mL
Boiling Point 203.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4OH
Refractive Index 1.5390 to 1.5420
Quantity 2.5 kg
Beilstein 06,373
Merck Index 15,2564
Specific Gravity 1.03
Solubility Information Solubility in water: in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Cresol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • m-Cresol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.