missing translation for 'onlineSavingsMsg'
Learn More

m-Cresol, 0.99%, Honeywell™

Product Code. p-7120550 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
100 g
250 g
2 kg
Packaging:
Glass bottle
Unit Size:
100g
250g
2kg
5g
This item is not returnable. View return policy

Product Code. 15677780

missing translation for 'mfr': Honeywell Research Chemicals C857275G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-39-4
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002302
InChI Key RLSSMJSEOOYNOY-UHFFFAOYSA-N
Synonym m-cresol, metacresol, meta-cresol, phenol, 3-methyl, 3-hydroxytoluene, m-methylphenol, 3-cresol, m-kresol, m-oxytoluene, m-toluol
PubChem CID 342
ChEBI CHEBI:17231
IUPAC Name 3-methylphenol
SMILES CC1=CC(=CC=C1)O

Specifications

Melting Point 11.8°C (1013hPa)
Color Colorless to Yellow
Density 1.034g/mL at 25°C
Boiling Point 202°C (1013 hPa)
Flash Point 86°C (186.8°F)
Ignition Point 1036°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 5 g
UN Number UN2076
Beilstein 506719
Solubility Information 22.7g/L at 25°C at ca. 7 pH Solubility in water
Vapor Density 3.72 (vs air)
Formula Weight 108.14g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material m-Cresol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.