missing translation for 'onlineSavingsMsg'
Learn More

m-Bromotoluene, MP Biomedicals

Product Code. 11438374 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438374 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438374 Supplier MP Biomedicals Supplier No. 0521077610
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 591-17-3
Molecular Formula C7H7Br
MDL Number MFCD00000085
Quantity 10 g
Synonym 3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
InChI Key WJIFKOVZNJTSGO-UHFFFAOYSA-N
SMILES CC1=CC=CC(Br)=C1
IUPAC Name 1-bromo-3-methylbenzene
Molecular Weight (g/mol) 171.04
PubChem CID 11560
Formula Weight 171.04
Chemical Name or Material m-Bromotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.