missing translation for 'onlineSavingsMsg'
Learn More

m-Anisidine, 98%

Product Code. 11469752
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469752 250 g 250g
11459752 50 g 50g
11449752 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469752 Supplier Thermo Scientific Alfa Aesar Supplier No. A10815.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 536-90-3 | C7H9NO | 123.155 g/mol

The unusually large amount of dibromo product produced upon bromination of m-anisidine may be attributed to the two doubly activeated positions. The best enantioselectivity of 97 % ee was observed for the reaction of m-anisidine in organocatalytic asymmetric three-component cyclization of cinnamaldehydes and primary amines with 1, 3-Dicarbonyl Compounds. Evidence for the control of 2nd-harmonic generation activities from the x-ray crystal-structures of the complexes of l-tartaric acid with m-anisidine and p-toluidine was determined.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The unusually large amount of dibromo product produced upon bromination of m-anisidine may be attributed to the two doubly activeated positions. The best enantioselectivity of 97 % ee was observed for the reaction of m-anisidine in organocatalytic asymmetric three-component cyclization of cinnamaldehydes and primary amines with 1, 3-Dicarbonyl Compounds. Evidence for the control of 2nd-harmonic generation activities from the x-ray crystal-structures of the complexes of l-tartaric acid with m-anisidine and p-toluidine was determined.

Solubility
Soluble in water(18g/L).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-90-3
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007783
InChI Key NCBZRJODKRCREW-UHFFFAOYSA-N
Synonym m-anisidine, 3-aminoanisole, m-methoxyaniline, 3-methoxybenzenamine, 3-anisidine, m-aminoanisole, benzenamine, 3-methoxy, m-anisylamine, 1-amino-3-methoxybenzene, unii-jxa144kx2i
PubChem CID 10824
IUPAC Name 3-methoxyaniline
SMILES COC1=CC=CC(=C1)N

Specifications

Melting Point -1°C
Density 1.101
Boiling Point 250°C to 251°C
Flash Point 126°C (258°F)
Odor Characteristic
Refractive Index 1.581
Quantity 250 g
UN Number UN2431
Beilstein 386119
Sensitivity Light sensitive
Merck Index 14,667
Solubility Information Soluble in water(18g/L).
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material m-Anisidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • m-Anisidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: m-Anisidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.