missing translation for 'onlineSavingsMsg'
Learn More

m-Anisic acid, 98%

Product Code. 15780809 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15780809 100 g Plastic bottle 100g
10512171 25 g Plastic bottle 25g
10380544 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15780809 Supplier Thermo Scientific Acros Supplier No. 152471000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 586-38-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-38-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002499
InChI Key XHQZJYCNDZAGLW-UHFFFAOYSA-N
Synonym m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425
PubChem CID 11461
IUPAC Name 3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 103.0°C to 108.0°C
Color Beige-Yellow to White
Boiling Point 170.0°C to 172.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3OC6H4CO2H
Quantity 100 g
Beilstein 10, 137
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material m-Anisic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.