missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Anisaldehyde, 97%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 591-31-1 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
InChI Key | WMPDAIZRQDCGFH-UHFFFAOYSA-N |
Synonym | m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh |
PubChem CID | 11569 |
IUPAC Name | 3-methoxybenzaldehyde |
SMILES | COC1=CC=CC(=C1)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10113210
|
Thermo Scientific Acros
156760250 |
25 mL | Glass bottle |
N/A
|
N/A | ||||
10275770
|
Thermo Scientific Acros
156761000 |
100 mL | Glass bottle |
N/A
|
N/A | ||||
11934791
|
Thermo Scientific Acros
156765000 |
500 mL | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
591-31-1 | |
136.15 | |
m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh | |
3-methoxybenzaldehyde |
C8H8O2 | |
WMPDAIZRQDCGFH-UHFFFAOYSA-N | |
11569 | |
COC1=CC=CC(=C1)C=O |
Specifications
591-31-1 | |
1.1190g/mL | |
110°C | |
96% min. (GC) | |
CH3OC6H4CHO | |
08,59 | |
m-anisaldehyde, 3-anisaldehyde, benzaldehyde, 3-methoxy, m-methoxybenzaldehyde, metamethoxybenzaldehyde, 3-methoxy-benzaldehyde, unii-8zao7s0ivh, ccris 960, meta-anisaldehyde, 8zao7s0ivh | |
WMPDAIZRQDCGFH-UHFFFAOYSA-N | |
3-methoxybenzaldehyde | |
11569 | |
97% | |
m-Anisaldehyde |
Yellow | |
143.0°C (50.0 mmHg) | |
Authentic | |
C8H8O2 | |
1.5500 to 1.5540 | |
1.119 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether | |
COC1=CC=CC(=C1)C=O | |
136.15 | |
136.15 | |
Liquid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-712-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- m-Anisaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only