missing translation for 'onlineSavingsMsg'
Learn More

m-Acetotoluidide, Thermo Scientific™

Product Code. 10187273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187273 25 g Glass bottle 25g
10493925 5 g Glass bottle 5g
10278703 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10187273 Supplier Thermo Scientific Acros Supplier No. 391300250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 537-92-8
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00014962
InChI Key ALMHSXDYCFOZQD-UHFFFAOYSA-N
Synonym 3'-methylacetanilide, n-acetyl-m-toluidine, m-acetotoluidide, 3-methylacetanilide, n-3-methylphenyl acetamide, m-acetotoluide, 3-acetamidotoluene, n-m-tolylacetamide, m-methylacetanilide, aceto-m-aminotoluene
PubChem CID 10843
IUPAC Name N-(3-methylphenyl)acetamide
SMILES CC1=CC(=CC=C1)NC(=O)C

Specifications

Melting Point 65°C to 67°C
Boiling Point 303°C
Packaging Glass bottle
Linear Formula CH3C6H4NHC(O)CH3
Quantity 25 g
Merck Index 15, 72
Formula Weight 149.19
Chemical Name or Material m-Acetotoluidide
compliance-icons
Product Identifier
  • m-Acetotoluidide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.