missing translation for 'onlineSavingsMsg'
Learn More

M-110, MedChemExpress

Product Code. 16465378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465378 10 mg 10mg
16458097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465378 Supplier Medchem Express Supplier No. HY12830
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress M-110 is a highly selective, ATP-competitive inhibitor of PIM kinases with a preference for PIM-3 (IC50=47 nM). M-110 inhibits PIM-1 and PIM-2 with similar IC50s of 2.5 μM. M-110 inhibits the proliferation of prostate cancer cell lines with IC50s of 0.6 to 0.9 μM.

M-110 is a highly selective, ATP-competitive inhibitor of PIM kinases with a preference for PIM-3 (IC50=47 nM). M-110 inhibits PIM-1 and PIM-2 with similar IC50s of 2.5 μM. M-110 inhibits the proliferation of prostate cancer cell lines with IC50s of 0.6 to 0.9 μM.

Specifications

Chemical Name or Material M-110
CAS 1395048-49-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H28ClN5O3
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (74.74 mM; Need ultrasonic)
SMILES O=C(N/N=C(C1=CC=C(Cl)C=C1O)\CC)C2=CC(NCCCN3CCOCC3)=NC=C2
Molecular Weight (g/mol) 445.94
Formula Weight 445.94
Percent Purity 95.27%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.