Learn More
LZTS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15677-1-AP-20UL
 View more versions of this product
Product Code. 16834954
  • £135.00 / 20µL
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 7p.
Specifications
Specifications
LZTS2
Polyclonal
Unconjugated
LZTS2
hLZTS2, KIAA1813, LAPSER1, LZTS2, Protein LAPSER1
Rabbit
Antigen Affinity Chromatography
RUO
226154, 365468, 84445
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3LUD4, Q91YU6, Q9BRK4
Lzts2
LZTS2 Fusion Protein Ag8145
20 μL
Primary
Rat, Human, Mouse