missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTS2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41099CL1
This item is not returnable.
View return policy
Description
LZTS2 Polyclonal antibody specifically detects LZTS2 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| LZTS2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| hLZTS2, KIAA1813leucine zipper putative tumor suppressor 2, LAPSER1Protein LAPSER1, leucine zipper, putative tumor suppressor 2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human LZTS2. The immunogen is located within amino acids 570 - 620 of LZTS2. Amino Acid Squence: LQRERRRGEEQRDSFEG | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 84445 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction