missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41099
This item is not returnable.
View return policy
Description
LZTS2 Polyclonal specifically detects LZTS2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LZTS2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LZTS2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human LZTS2. The immunogen is located within amino acids 570 - 620 of LZTS2. Amino Acid Squence: LQRERRRGEEQRDSFEG | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 84445 | |
| Human | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| hLZTS2, KIAA1813leucine zipper putative tumor suppressor 2, LAPSER1Protein LAPSER1, leucine zipper, putative tumor suppressor 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| LZTS2 antibody is predicted to not cross-react with other LZTS family members. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction