missing translation for 'onlineSavingsMsg'
Learn More

LZTS2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41152-0.025mg

 View more versions of this product

Product Code. 18634496

  • £134.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LZTS2 Polyclonal specifically detects LZTS2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LZTS2
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
LZTS2
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human LZTS2. The immunogen is located within amino acids 80 - 130 of LZTS2. Amino Acid Squence: INEDFRTESPPSPSSDVED
0.025 mg
Cell Cycle and Replication
84445
Human, Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
hLZTS2, KIAA1813leucine zipper putative tumor suppressor 2, LAPSER1Protein LAPSER1, leucine zipper, putative tumor suppressor 2
Rabbit
Affinity Purified
RUO
Primary
LZTS2 antibody is predicted to not cross-react with other LZTS family members
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.