missing translation for 'onlineSavingsMsg'
Learn More

LZTS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18373017 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373017 20 μL 20µL
18333054 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18373017 Supplier Novus Biologicals Supplier No. NBP30496920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LZTS1 Polyclonal specifically detects LZTS1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias F37, F37/esophageal cancer-related gene-coding leucine-zipper motifFez1, FEZ1leucine zipper putative tumor suppressor 1, leucine zipper, putative tumor suppressor 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 337-596 of human LZTS1 (NP_066300.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11178
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.