Learn More
LZTS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541614
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QQSYVAMYQR NQRLEKALQQ LARGDSAGEP LEVDLEGADI PYEDIIATEI Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
LZTS1 Polyclonal antibody specifically detects LZTS1 in Human samples. It is validated for Western Blot.Specifications
LZTS1 | |
Polyclonal | |
Unconjugated | |
Lzts1 | |
F37; F37 esophageal cancer-related leucine zipper motif 1; F37/Esophageal cancer-related gene-coding leucine-zipper motif; FEZ1; leucine zipper putative tumor suppressor 1; leucine zipper tumor suppressor 1; leucine zipper, putative tumor suppressor 1; leucine-zipper protein FEZ1; LOW QUALITY PROTEIN: leucine zipper putative tumor suppressor 1; LZTS1; Psdzip70; PSD-Zip70 | |
Rabbit | |
Protein A | |
RUO | |
11178 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y250 | |
Lzts1 | |
Synthetic peptide directed towards the C-terminal of human LZTS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |