missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTR2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77122CL1
This item is not returnable.
View return policy
Description
LZTR2 Polyclonal antibody specifically detects LZTR2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| LZTR2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| DKFZp686C2486, FLJ23871, FLJ25761, KIAA1928, Leucine zipper transcription regulator 2FLJ36620, LZTR2SEC16Sprotein SEC16 homolog B, PGPR-p117, protein transport protein Sec16B, Regucalcin gene promoter region-related protein p117, regucalcin gene promotor region related protein, RGPRFLJ33652, RGPR-p117, SEC16 homolog B, SEC16 homolog B (S. cerevisiae) | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human LZTR2. The immunogen is located within the last 50 amino acids of LZTR2. Amino Acid Squence: TSLNRPNRLAQRRYPTQP | |
| 0.1 mL | |
| metabolism | |
| 89866 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction