Learn More
Invitrogen™ LZTR1 Recombinant Rabbit Monoclonal Antibody (1P0U1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557965
Description
Immunogen sequence: GGAADNTLPN ELHCYDVDFQ TWEVVQPSSD SEVGGAEVPE RACASEEVPT LTYEERVGFK KSRDVFGLDF GTTSAKQPTQ PASELPSGRL FHAAAVISDA MYIFGGTVDN NIRSGEMYRF QFSCYPKCTL HEDYGRLWES RQFCDVEFVL GEKEECVQGH VAIVTARSRW LRRKITQARE RLAQKLEQEA APVPREAPGV AAGGARPPLL HVAIREAEAR PFEVLMQFLY TDKIKYPRKG HVEDVL.
LZTR1, a member of the BTB-kelch superfamily, was initially described as a putative transcriptional regulator based on weak homology to members of the basic leucine zipper-like family, the encoded protein subsequently has been shown to localize exclusively to the Golgi network where it may help stabilize the Golgi complex. Deletion of this gene may be associated with DiGeorge syndrome, a developmental field defect involving the third and fourth pharyngeal pouches, causing the absence of thymus and parathyroid glands, congenital cardiac abnormalities and facial dysmorphism. LZTR1 is tyrosine phosphorylated and subsequently degraded upon induction of apoptosis.
Specifications
| LZTR1 | |
| Recombinant Monoclonal | |
| 1.66 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9CQ33 | |
| LZTR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-545 of human LZTR1 (NP_006758.2). | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1P0U1 | |
| Unconjugated | |
| LZTR1 | |
| 1200003E21Rik; AI591627; AW550890; BTBD29; fj87c05; leucine zipper like transcription regulator 1; leucine-zipper-like transcription regulator 1; leucine-zipper-like transcriptional regulator 1; leucine-zipper-like transcriptional regulator, 1; Lztr1; LZTR-1; MGC21205; NS10; si:dkey-98f17.2; SWNTS2; Tcfl2; wu:fj87c05; zgc:158250 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 66863 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.