missing translation for 'onlineSavingsMsg'
Learn More

LZTR1 Polyclonal Antibody

Product Code. 12849042
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849042 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849042 Supplier Invitrogen Supplier No. PA520902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LZTR1 Polyclonal Antibody

LZTR1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen LZTR1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LZTR1
Gene Accession No. Q8N653, Q9CQ33
Gene Alias 1200003E21Rik; AI591627; AW550890; BTBD29; fj87c05; leucine zipper like transcription regulator 1; leucine-zipper-like transcription regulator 1; leucine-zipper-like transcriptional regulator 1; leucine-zipper-like transcriptional regulator, 1; Lztr1; LZTR-1; MGC21205; NS10; si:dkey-98f17.2; SWNTS2; Tcfl2; wu:fj87c05; zgc:158250
Gene Symbols LZTR1
Host Species Rabbit
Immunogen A 14 amino acid peptide near the amino terminus of human LZTR1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360745, 66863, 8216
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.