missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LZTR1 Polyclonal antibody specifically detects LZTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | LZTR1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | leucine-zipper-like regulator-1, leucine-zipper-like transcription regulator 1, leucine-zipper-like transcriptional regulator 1, LZTR-1, MGC21205, TCFL2 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human LZTR1. The immunogen is located within amino acids 130 - 180 of LZTR1. Amino Acid Squence: NSNLKNKNDLFEYK |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?