Learn More
LZTFL1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2C1F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68281-20UL
Description
The LZTFL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LZTFL1. This antibody recognizes human antigen. The LZTFL1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| LZTFL1 | |
| 2C1F6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| FLJ36386, LZTFL1 | |
| 20 μL | |
| 54585 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |