missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYVE-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 19 publications
£190.00 - £492.00
Specifications
| Antigen | LYVE-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1-2 ug/mL, Flow Cytometry 3-20 ug/mL, ELISA 1-15 ug/mL, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:800, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating, Immunofluorescence, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunofluorescence, CyTOF |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18011384
|
Novus Biologicals
NB600-1008SS |
0.025 mg |
£190.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706053
|
Novus Biologicals
NB600-1008 |
0.2 mg |
£492.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYVE-1 Polyclonal specifically detects LYVE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunofluorescence, CyTOF-ready.Specifications
| LYVE-1 | |
| Western Blot 1-2 ug/mL, Flow Cytometry 3-20 ug/mL, ELISA 1-15 ug/mL, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:800, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating, Immunofluorescence, CyTOF-ready | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q8BHC0 | |
| 10894 | |
| Produced from sera of rabbits immunized with highly pure recombinant mouse soluble LYVE-1 produced in insect cells. The recombinant soluble LYVE-1 consists of amino acid 24 (Ala) to 228 (Gly) and is fused to a C-terminal His-tag (6xHis). | |
| Primary | |
| Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. Please note the sample size of this product will be provided in reconstituted liquid form. | |
| 35 kDa |
| LYOPH | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunofluorescence, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
| PBS with No Preservative | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE-1XLKD1 | |
| LYVE1 | |
| IgG | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title