missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Antibody (simtuzumab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28051JF525
This item is not returnable.
View return policy
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Lysyl Oxidase Homolog 2/LOXL2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
| LOXL2 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4017 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| simtuzumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction