missing translation for 'onlineSavingsMsg'
Learn More

Lysyl Oxidase Homolog 2/LOXL2 Antibody (OTI8B2), DyLight 594, Novus Biologicals™

Product Code. 18413566 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413566 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413566 Supplier Novus Biologicals Supplier No. NBP272558DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysyl Oxidase Homolog 2/LOXL2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI8B2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 152-450 of human Lysyl Oxidase Homolog 2/LOXL2 (NP_002309) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4017
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.