missing translation for 'onlineSavingsMsg'
Learn More

Lysozyme/Muramidase Ab-1 Rabbit Polyclonal Antibody, Epredia™

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Lysozyme/Muramidase Ab-1, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-372-A

 View more versions of this product

Product Code. 12603267

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Lysozyme is synthesized predominantly in reactive histiocytes rather than in resting, unstimulated phagocytes.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey, Pig, Cat and Dog. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Human lysozyme purified from the urine of patients with monocytic leukemia

Positive Control: Tonsil, colon or skin

Cellular Localization: Cytoplasmic

Ab-1 was adsorbed with human plasma and urine proteins to remove the cross-reacting antibodies. Ab-1 labels myeloid cells, histiocytes, granulocytes, macrophages, and monocytes. This antibody is helpful in the identification of myeloid or monocytic nature of acute leukemia.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lysozyme/Muramidase Ab-1
Polyclonal
Unconjugated
Rabbit
1 mL
Enzyme
Canine, Feline, Human, Non-human Primate
Concentrated
Immunohistochemistry (Paraffin)
Polyclonal
1:100-1:200 for 30 min at RT
Human lysozyme purified from the urine of patients with monocytic leukemia
IVD
Primary
Store vial at 4deg.C. When stored at 2 to 8deg.C,
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.