Learn More
Invitrogen™ Lysozyme Monoclonal Antibody (LZ-2), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ GIC207
Product Code. 10456243
  • £204.00 / 1mL
Edge
For Research Use Only. All usage must comply with product instructions.
Description
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
This gene encodes human lysozyme, whose natural substrate is the bacterial cell wall peptidoglycan (cleaving the beta[1-4]glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine). Lysozyme is one of the anti-microbial agents found in human milk, and is also present in spleen, lung, kidney, white blood cells, plasma, saliva, and tears. Missense mutations in LYZ have been identified in heritable renal amyloidosis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lysozyme
Monoclonal
FITC
LYZ
1,4-beta-N-acetylmuramidase C; 1700038F02Rik; 4-beta-N-acetylmuramidase C; AI326280; Allergen Gal d IV; bA534G20.1; c-type lysozyme; dystrophin; egg white lysozyme; Gal d 4; KAAG648; LYC2; Lys; Lysm; lysozyme; lysozyme (renal amyloidosis); lysozyme 1; lysozyme 2; lysozyme C; Lysozyme C type M; lysozyme C type P; Lysozyme C, spleen isozyme; lysozyme C-1; Lysozyme C-2; lysozyme C-3; lysozyme d1; lysozyme F1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; Lysz; LYZ; Lyz1; Lyz2; LYZC; LYZD1; LYZF1; LYZL1; Lyzs; Lzm; Lzm-s1; Lzp; Lzp-s; P lysozyme structural; PRO1278; renal amyloidosis; RGD1559869; unnamed protein product; UNQ648/PRO1278
Mouse
Purified
ASR
4069
4°C, store in dark
Liquid
Responsibly packaged
Flow Cytometry
LZ-2
PBS with 1% BSA and 0.05% sodium azide; pH 7.2
P61626
LYZ
Human Lysozyme.
1 mL
Primary
Human
Antibody
IgG1