missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Lysozyme (Egg White)

Product Code. 16111780
Click to view available options
:
1g
This item is not returnable. View return policy

Product Code. 16111780

Brand: Abnova™ P5237.1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

Accession Number P00698
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 396218
Molecular Weight (g/mol) 14.3kDa
Name Lysozyme (Egg White)
Preparation Method Native protein purified from Egg Whites
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Gene Alias LYZC
Common Name LYZ
Gene Symbol LYZ
Assays Lysozyme Assay; Nucleic acid preparation; Protein purification from inclusion bodies; Plasmid preparation (to break down membranes and cell wall); Hydrolysis of chitin, Lysozyme Assay; Hydrolysis of bacterial cell walls.
Biological Activity ≥5,000 units per mg dry weight. One Unit is equal to a decrease in turbidity of 0.001 per minute at 450nm at pH 7.0 and 25°C, using a 0.3mg/ml suspension of Micrococcus lysodeikticus cells as substrate.
Species Chicken
Protein Tag None
Expression System Native protein purified from Egg Whites
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.