missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Lysozyme (Egg White), 2X
Click to view available options
:
1g
Description
#REF!
Specifications
Specifications
| Accession Number | P00698 |
| For Use With (Application) | Enzyme Activity |
| Gene ID (Entrez) | 396218 |
| Molecular Weight (g/mol) | 14.3kDa |
| Name | Lysozyme (Egg White), 2x |
| Preparation Method | Native protein purified from Egg Whites |
| Quantity | 1 g |
| Storage Requirements | Store at 4°C. |
| Regulatory Status | RUO |
| Gene Alias | LYZC |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction