missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysozyme Antibody (LZ598-10G9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62223AF405
This item is not returnable.
View return policy
Description
Lysozyme Monoclonal specifically detects Lysozyme in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| Lysozyme | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LYZ | |
| human lysozyme | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-188 reacts with a 180 kDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that the antibody MEM-188 could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunocytochemistry | |
| LZ598-10G9 | |
| Flow Cytometry, Immunocytochemistry | |
| EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis | |
| Mouse | |
| Protein A purified | |
| Cardiovascular Biology, Immunology | |
| 4069 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction